missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Methyl-1-pentene, TCI America™

Manufacturer:  TCI America M05755G

 View more versions of this product

Catalog No. M05755G



Specifications

Specifications

3-Methyl-1-pentene
Colorless
760-20-3
MFCD00009339
3-methyl-1-pentene, 1-pentene, 3-methyl, ethene, trimer, 2-vinylbutane, sec-butylethene, 3-methylpentene-1, pentene, 3-methyl, 1-pentene,3-methyl, acmc-1bmfr, 3-methyl-1-pentene
CCC(C)C=C
84.162
84.16
≥98.0% (GC)
54°C
5g
C6H12
2288
LDTAOIUHUHHCMU-UHFFFAOYSA-N
3-methylpent-1-ene
12969
Clear Liquid at 20°C

Chemical Identifiers

760-20-3
84.162
LDTAOIUHUHHCMU-UHFFFAOYSA-N
12969
CCC(C)C=C
C6H12
MFCD00009339
3-methyl-1-pentene, 1-pentene, 3-methyl, ethene, trimer, 2-vinylbutane, sec-butylethene, 3-methylpentene-1, pentene, 3-methyl, 1-pentene,3-methyl, acmc-1bmfr, 3-methyl-1-pentene
3-methylpent-1-ene
Safety and Handling

Safety and Handling

EINECSNumber : (2)-0031

TSCA : No

Documents