Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Methyl-1-penten-3-ol, 98%

Chemical Identifiers

Chemical Name or Material 3-Methyl-1-penten-3-ol
CAS 918-85-4
Molecular Formula C6H12O
Formula Weight 100.16
MDL Number MFCD00004481
UN Number UN1987
Synonym Ethyl methyl vinyl carbinol
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2089306

 
alfa aesar™
B2089306
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAB2089314

 
alfa aesar™
B2089314
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 100.16
MDL Number MFCD00004481
CAS 918-85-4
RTECS Number SB3495000
Boiling Point 118°C
Chemical Name or Material 3-Methyl-1-penten-3-ol
TSCA TSCA
Assay 98%
Molecular Formula C6H12O
UN Number UN1987
Synonym Ethyl methyl vinyl carbinol
Flash Point 26°C (79°F)
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301-H226
Toxic if swallowed.
Flammable liquid and vapour.
Health Hazard 3 GHS P Statement
P210-P280-P301+P310-P303+P361+P353-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ALCOHOLS, N.O.S.
Quantity 5g

Applications
It is employed in the optimized palladium catalyst systems for the selective liquid-phase hydrogenation of functionalized alkynes and in the bacterial degradation of tert-amyl alcohol proceeds via hemiterpene 2-methyl-3-buten-2-ol by employing the tertiary alcohol desaturase function of the rieske nonheme mononuclear iron oxygenase MdpJ.

Solubility
Not miscible or difficult to mix in water. Soluble in alcohol.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Keep away from ignition sources. Protect from electrostatic charges.

Safety and Handling

Danger

GHS P Statement
P210-P280-P301+P310-P303+P361+P353-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H226
Toxic if swallowed.
Flammable liquid and vapour.