Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Methyl-1-nonyn-3-ol, 97%

$39.32 - $421.89

Chemical Identifiers

Chemical Name or Material 3-Methyl-1-nonyn-3-ol
CAS 5430-01-3
Molecular Formula C10H18O
Formula Weight 154.25
MDL Number MFCD00041582
Beilstein 1750318
Synonym Ethynyl n-hexyl methyl carbinol
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2410606

  SDS
alfa aesar™
B2410606
5g Each for $39.32

AAB2410614

  SDS
alfa aesar™
B2410614
25g Each for $135.61

AAB2410622

  SDS
alfa aesar™
B2410622
100g Each for $421.89
Description & Specifications

Specifications

Formula Weight 154.25
CAS 5430-01-3
Quantity 5g
Assay 97%
Boiling Point 87° to 88°C (15mmHg)
Molecular Formula C10H18O
Synonym Ethynyl n-hexyl methyl carbinol
Chemical Name or Material 3-Methyl-1-nonyn-3-ol
Beilstein 1750318
MDL Number MFCD00041582