missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Methyl-1-hexene, TCI America™

$149.72 - $441.53

Chemical Identifiers

CAS 3404-61-3
Molecular Formula C7H14
Molecular Weight (g/mol) 98.189
MDL Number MFCD00009407
InChI Key RITONZMLZWYPHW-UHFFFAOYSA-N
PubChem CID 18848
IUPAC Name 3-methylhex-1-ene
SMILES CCCC(C)C=C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M03011ML View Documents TCI America
M03011ML
1mL Each for $149.72
 
M03015ML View Documents TCI America
M03015ML
5mL Each for $441.53
 
Specifications

Chemical Identifiers

3404-61-3
98.189
RITONZMLZWYPHW-UHFFFAOYSA-N
3-methylhex-1-ene
C7H14
MFCD00009407
18848
CCCC(C)C=C

Specifications

84°C
1mL
C7H14
3295
CCCC(C)C=C
98.189
98.19
≥99.0% (GC)
Colorless
3404-61-3
MFCD00009407
RITONZMLZWYPHW-UHFFFAOYSA-N
3-methylhex-1-ene
18848
Clear Liquid at 20°C
3-Methyl-1-hexene
Safety and Handling

Safety and Handling

TSCA : Yes