missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Methyl-1-hexene 99.0+%, TCI America™

Manufacturer:  TCI America M03011ML

 View more versions of this product

Catalog No. M03011ML



Specifications

Specifications

3-Methyl-1-hexene
Colorless
3404-61-3
MFCD00009407
RITONZMLZWYPHW-UHFFFAOYSA-N
3-methylhex-1-ene
18848
Clear Liquid at 20°C
84°C
1mL
C7H14
3295
CCCC(C)C=C
98.189
98.19
≥99.0% (GC)

Chemical Identifiers

3404-61-3
98.189
RITONZMLZWYPHW-UHFFFAOYSA-N
3-methylhex-1-ene
C7H14
MFCD00009407
18848
CCCC(C)C=C
Safety and Handling

Safety and Handling

TSCA : Yes

Documents