missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Methyl-1-cyclohexene, 90%, Tech., Thermo Scientific Chemicals

 

Supplier:  Thermo Scientific Chemicals 304310250

Catalog No. AC304310250



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

3-Methyl-1-cyclohexene
591-48-0
100.0
0.8000g/mL
-3°C
88% min. (GC)
C7H12
C6H9CH3
25 mL
0.8
UZPWKTCMUADILM-UHFFFAOYSA-N
3-methylcyclohexene
11573
90%
Liquid
tech., 90%
88.0
Colorless
104°C
Authentic
Glass bottle
1.4425 to 1.4445
MFCD00001571
05, 67
3-methyl-1-cyclohexene, cyclohexene, 3-methyl, 3-methyl cyclohexene, 3-methyl-cyclohexene, 3-methylcyclohexene-1, 3-methylcyclohex-1-ene, acmc-1atr1
CC1CCCC=C1
96.17
96.17
Technical

Chemical Identifiers

591-48-0
96.17
UZPWKTCMUADILM-UHFFFAOYSA-N
11573
CC1CCCC=C1
C7H12
MFCD00001571
3-methyl-1-cyclohexene, cyclohexene, 3-methyl, 3-methyl cyclohexene, 3-methyl-cyclohexene, 3-methylcyclohexene-1, 3-methylcyclohex-1-ene, acmc-1atr1
3-methylcyclohexene
Safety and Handling

Safety and Handling

GHS H Statement:
Highly flammable liquid and vapor.

GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 209-717-5

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only