Please login to your online account to display your discounted pricing

3-Methyl-1-butyne 96%, ACROS Organics™


Quantity 10mL
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC392740100

 View more versions of this product

Catalog No. AC392740100

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥95.0%
Formula Weight 68.12
Physical Form Liquid
Quantity 10mL
Color Clear colorless to slightly yellow
Density 0.666g/mL
CAS Max % 100.0
CAS Min % 95.0
Synonym Isopropylacetylene
Infrared Spectrum Authentic
Chemical Name or Material 3-Methyl-1-butyne
Assay Percent Range 96%
Health Hazard 1 GHS Signal Word: Danger
MDL Number MFCD00039853
Refractive Index 1.3730 to 1.3750 (20°C, 589nm)
Packaging Glass bottle
Linear Formula (CH3)2CHC2CH
Molecular Formula C5H8
CAS 598-23-2
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.
Take precautionary measures against static discharge.
Store in a well-ventilated place. Keep cool.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Extremely flammable liquid and vapor.

Specifications

Chemical Name or Material 3-Methyl-1-butyne
CAS 598-23-2
CAS Min % 95.0
CAS Max % 100.0
Assay ≥95.0%
Assay Percent Range 96%
Molecular Formula C5H8
Linear Formula (CH3)2CHC2CH
Formula Weight 68.12
MDL Number MFCD00039853
Synonym Isopropylacetylene
Accessories

GHS Signal Word: Danger

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.
Take precautionary measures against static discharge.
Store in a well-ventilated place. Keep cool.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Extremely flammable liquid and vapor.

SDS & Safety Documents