Please login to your online account to display your discounted pricing

3-Methyl-1-butyne 96%, ACROS Organics™


Quantity 1mL
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC392740010

 View more versions of this product

Catalog No. AC392740010

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥95.0%
Physical Form Liquid
Formula Weight 68.12
Quantity 1mL
Color Clear colorless to slightly yellow
Density 0.666g/mL
CAS Max % 100.0
CAS Min % 95.0
Linear Formula (CH3)2CHC2CH
Infrared Spectrum Authentic
CAS 598-23-2
Assay Percent Range 96%
Chemical Name or Material 3-Methyl-1-butyne
Health Hazard 1 GHS Signal Word: Danger
MDL Number MFCD00039853
Packaging Glass bottle
Synonym Isopropylacetylene
Molecular Formula C5H8
Refractive Index 1.3730 to 1.3750 (20°C, 589nm)
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Extremely flammable liquid and vapor.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.
Take precautionary measures against static discharge.
Store in a well-ventilated place. Keep cool.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material 3-Methyl-1-butyne
CAS 598-23-2
CAS Min % 95.0
CAS Max % 100.0
Assay ≥95.0%
Assay Percent Range 96%
Molecular Formula C5H8
Linear Formula (CH3)2CHC2CH
Formula Weight 68.12
MDL Number MFCD00039853
Synonym Isopropylacetylene
Accessories

GHS Signal Word: Danger

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.
Take precautionary measures against static discharge.
Store in a well-ventilated place. Keep cool.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Extremely flammable liquid and vapor.

SDS & Safety Documents