Learn More
3-Methyl-1-butanol, ACS reagent, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 412735000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
3-Methyl-1-butanol | |
123-51-3 | |
0.8000g/mL | |
45°C | |
C5H12O | |
(CH3)2CHCH2CH2OH | |
500 mL | |
0.003% max. | |
15, 5241 | |
Solubility in water: 25g/L (20°C). Other solubilities: miscible with alcohol,ether,benzene,oils,,chloroform | |
CC(C)CCO | |
88.15 | |
4.3 mPa.s (20°C) | |
88.15 | |
ACS Reagent | |
Liquid |
98.5% | |
-70.0°C | |
130.0°C | |
Glass bottle | |
1.4050 to 1.4070 | |
MFCD00002934 | |
01, 392 | |
0.8 | |
3-methyl-1-butanol, isoamyl alcohol, isopentyl alcohol, isopentanol, 3-methylbutanol, 1-butanol, 3-methyl, isoamylol, isobutylcarbinol, iso-amylalkohol, isobutyl carbinol | |
PHTQWCKDNZKARW-UHFFFAOYSA-N | |
3-methylbutan-1-ol | |
31260 | |
CHEBI:15837 | |
≥98.5% | |
0.002 meq/g max |
Chemical Identifiers
123-51-3 | |
88.15 | |
PHTQWCKDNZKARW-UHFFFAOYSA-N | |
31260 | |
3-methylbutan-1-ol |
C5H12O | |
MFCD00002934 | |
3-methyl-1-butanol, isoamyl alcohol, isopentyl alcohol, isopentanol, 3-methylbutanol, 1-butanol, 3-methyl, isoamylol, isobutylcarbinol, iso-amylalkohol, isobutyl carbinol | |
CHEBI:15837 | |
CC(C)CCO |
Safety and Handling
GHS H Statement
Flammable liquid and vapour.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Repeated exposure may cause skin dryness or cracking.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Wash with plenty of soap and water.
If skin
GHS Signal Word: Warning
EINECSNumber : 204-633-5
RTECSNumber : EL5425000
TSCA : TSCA
RUO – Research Use Only