Please login to your online account to display your discounted pricing

3-Methyl-1-butanol, 99%, anhydrous, AcroSeal™, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 3-Methyl-1-butanol
CAS 123-51-3
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5% (GC)
Assay Percent Range 99%
Molecular Formula C5H12O
Linear Formula (CH3)2CHCH2CH2OH
Formula Weight 88.15
Synonym Isoamyl alcohol
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC433871000

  SDS
thermo fisher scientific
AC433871000
AcroSeal™ Glass Bottle 100mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC433870010

  SDS
thermo fisher scientific
AC433870010
AcroSeal™ Glass Bottle 1L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Liquid
Assay ≥98.5% (GC)
Formula Weight 88.15
Quantity 100mL
Color Clear colorless
Packaging AcroSeal™ Glass Bottle
Density 0.8000g/mL
Water 0.005% Max.
CAS Max % 100.0
Refractive Index 1.4050 to 1.4070 (20°C, 589nm)
Residue after Evaporation 0.0005 % max.
Linear Formula (CH3)2CHCH2CH2OH
Synonym Isoamyl alcohol
Chemical Name or Material 3-Methyl-1-butanol
CAS 123-51-3
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement:
May cause respiratory irritation.
Harmful if inhaled.
Flammable liquid and vapor.
Repeated exposure may cause skin dryness or cracking.
Infrared Spectrum Authentic
Assay Percent Range 99%
Molecular Formula C5H12O
CAS Min % 98.5
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Safety and Handling

Warning

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause respiratory irritation.
Harmful if inhaled.
Flammable liquid and vapor.
Repeated exposure may cause skin dryness or cracking.