missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Methyl-1-butanethiol 96%, ACROS Organics™

Manufacturer:  Acros Organics 152051000

 View more versions of this product

Catalog No. AC152051000



Specifications

Specifications

3-Methyl-1-butanethiol
19°C
Glass bottle
0.835
Colorless to Yellow
96%
95.0
96%
(CH3)2CHCH2CH2SH
01, 405
isoamyl mercaptan, 3-methyl-1-butanethiol, 1-butanethiol, 3-methyl, isopentyl mercaptan, 3-methylbutanethiol, isoamylthiol, isopentanethiol, isoamylmercaptan, unii-mmk4sun45e, isoamyl sulfhydrate
CC(C)CCS
104.211
104.21
96%
0.8350g/mL
Authentic
1.4420 to 1.4460
120.0°C
100g
541-31-1
100.0
C5H12S
MFCD00004899
09, 4970
GIJGXNFNUUFEGH-UHFFFAOYSA-N
3-methylbutane-1-thiol
10925
Liquid

Chemical Identifiers

541-31-1
104.211
GIJGXNFNUUFEGH-UHFFFAOYSA-N
10925
CC(C)CCS
C5H12S
MFCD00004899
isoamyl mercaptan, 3-methyl-1-butanethiol, 1-butanethiol, 3-methyl, isopentyl mercaptan, 3-methylbutanethiol, isoamylthiol, isopentanethiol, isoamylmercaptan, unii-mmk4sun45e, isoamyl sulfhydrate
3-methylbutane-1-thiol
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if inhaled.
Causes skin irritation.
Harmful in contact with skin.
Harmful if swallowed.
Highly flammable liquid and vapor.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 208-774-3

TSCA : TSCA

SDS Documents