Please login to your online account to display your discounted pricing

3-Methyl-1-butanethiol 96%, ACROS Organics™

$40.53 - $95.06

Chemical Identifiers

Chemical Name or Material 3-Methyl-1-butanethiol
Name Note 96%
CAS 541-31-1
CAS Min % 95.0
CAS Max % 100.0
Assay ≥95%
Assay Percent Range 96%
Molecular Formula C5H12S
Linear Formula (CH3)2CHCH2CH2SH
Formula Weight 104.21
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC152050050

  SDS
thermo fisher scientific
AC152050050
Glass bottle 5g Each for $40.53

AC152050250

  SDS
thermo fisher scientific
152050250
Glass bottle 25g Each for $95.06

AC152051000

  SDS
thermo fisher scientific
152051000
Glass bottle 100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥95%
Physical Form Liquid
Formula Weight 104.21
Quantity 5g
Color Clear colorless to slightly yellow
Density 0.835g/mL
EINECS Number 208-774-3
TSCA TSCA
Beilstein 01, 405
Merck Index 09, 4970
CAS Min % 95.0
CAS Max % 100.0
Chemical Name or Material 3-Methyl-1-butanethiol
Infrared Spectrum Authentic
Name Note 96%
Packaging Glass bottle
Refractive Index 1.4420 to 1.4460 (20°C, 589nm)
Linear Formula (CH3)2CHCH2CH2SH
MDL Number MFCD00004899
Synonym Isoamyl mercaptan
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if inhaled.
Causes skin irritation.
Harmful in contact with skin.
Harmful if swallowed.
Highly flammable liquid and vapor.
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Assay Percent Range 96%
Molecular Formula C5H12S
CAS 541-31-1
Safety and Handling

GHS Signal Word: Danger

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if inhaled.
Causes skin irritation.
Harmful in contact with skin.
Harmful if swallowed.
Highly flammable liquid and vapor.