missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Methyl-1,5-pentanediol, TCI America™

Manufacturer:  TCI America M071025ML

 View more versions of this product

Catalog No. M071025ML



Specifications

Specifications

3-Methyl-1,5-pentanediol
Colorless
4457-71-0
MFCD00039547
SXFJDZNJHVPHPH-UHFFFAOYSA-N
3-methylpentane-1,5-diol
20524
Clear Liquid at 20°C
150°C
25mL
C6H14O2
3-methyl-1,5-pentanediol, 1,5-pentanediol, 3-methyl, 1,5-dihydroxy-3-methylpentane, unii-88495m4hns, acmc-1ai92, 3-methyl-1,5-pentane diol, 3-methyl-pentane-1,5-diol, 3-methylpentane-1,5-diol, 1,5-pentanediol,3-methyl, 4-01-00-02571 beilstein handbook reference
CC(CCO)CCO
118.176
118.18
≥97.0% (GC)

Chemical Identifiers

4457-71-0
118.176
SXFJDZNJHVPHPH-UHFFFAOYSA-N
20524
CC(CCO)CCO
C6H14O2
MFCD00039547
3-methyl-1,5-pentanediol, 1,5-pentanediol, 3-methyl, 1,5-dihydroxy-3-methylpentane, unii-88495m4hns, acmc-1ai92, 3-methyl-1,5-pentane diol, 3-methyl-pentane-1,5-diol, 3-methylpentane-1,5-diol, 1,5-pentanediol,3-methyl, 4-01-00-02571 beilstein handbook reference
3-methylpentane-1,5-diol
Safety and Handling

Safety and Handling

EINECSNumber : (2)-0240

RTECSNumber : SA0800000

TSCA : Yes

Documents