missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Methyl-1,3-butanediol 98.0+%, TCI America™

Manufacturer:  TCI America M0818500ML

 View more versions of this product

Catalog No. M0818500ML



Specifications

Specifications

3-Methyl-1,3-butanediol
Colorless
2568-33-4
MFCD00059655
XPFCZYUVICHKDS-UHFFFAOYSA-N
3-methylbutane-1,3-diol
247470
Clear Liquid at 20°C
203°C
500mL
C5H12O2
3-methyl-1,3-butanediol, 1,3-butanediol, 3-methyl, isopentyldiol, 2-methyl-2,4-butanediol, unii-19nol5474q, 2,4-dihydroxy-2-methylbutane, 3-methyl-butane-1,3-diol, 1,3-butanediol,3-methyl, 3-methyl-1,3-butandiol, 3-methylbutan-1,3-diol
CC(C)(CCO)O
104.149
104.15
≥98.0% (GC)

Chemical Identifiers

2568-33-4
104.149
XPFCZYUVICHKDS-UHFFFAOYSA-N
247470
CC(C)(CCO)O
C5H12O2
MFCD00059655
3-methyl-1,3-butanediol, 1,3-butanediol, 3-methyl, isopentyldiol, 2-methyl-2,4-butanediol, unii-19nol5474q, 2,4-dihydroxy-2-methylbutane, 3-methyl-butane-1,3-diol, 1,3-butanediol,3-methyl, 3-methyl-1,3-butandiol, 3-methylbutan-1,3-diol
3-methylbutane-1,3-diol
Safety and Handling

Safety and Handling

EINECSNumber : (2)-0240

TSCA : Yes

Documents