Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Methyl-1-(2-phenylethyl)-4-piperidone, 97%

$111.59 - $332.49

Chemical Identifiers

Chemical Name or Material 3-Methyl-1-(2-phenylethyl)-4-piperidone
CAS 129164-39-2
Molecular Formula C14H19NO
Formula Weight 217.32
MDL Number MFCD01699280
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2753203

 
alfa aesar™
H2753203
1g Each for $111.59

AAH2753206

 
alfa aesar™
H2753206
5g Each for $332.49
Description & Specifications

Specifications

Formula Weight 217.32
Chemical Name or Material 3-Methyl-1-(2-phenylethyl)-4-piperidone
Assay 97%
Molecular Formula C14H19NO
MDL Number MFCD01699280
CAS 129164-39-2
Quantity 1g

Applications
As intermediates in the synthesis of several piperidine derivatives. 4-Piperidone is used as an intermediate in the manufacture of chemicals and pharmaceutical drugs.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.