Please login to your online account to display your discounted pricing

3-Methyl-1,2-dihydropyridin-2-one, 97%, Maybridge


Quantity 5g
Packaging Amber glass bottle

Manufacturer: fisher scientific company  CD09405DE

 View more versions of this product

Catalog No. CD09405DE

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 109.13
Quantity 5g
Assay 97%
Chemical Name or Material 3-Methyl-1, 2-dihydropyridin-2-one
Assay Percent Range 97%
Molecular Formula C6H7NO
CAS 1003-56-1
Packaging Amber glass bottle

Specifications

Chemical Name or Material 3-Methyl-1, 2-dihydropyridin-2-one
Assay 97%
Assay Percent Range 97%
Molecular Formula C6H7NO
Formula Weight 109.13