Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Methyl-1,2-cyclopentanediol, mixture of diastereomers, 95%

$37.36 - $580.11

Chemical Identifiers

Chemical Name or Material 3-Methyl-1,2-cyclopentanediol
CAS 27583-37-5
Name Note mixture of diastereomers
Molecular Formula C6H12O2
Formula Weight 116.16
MDL Number MFCD00019285
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2044003

  SDS
alfa aesar™
B2044003
1g Each for $37.36

AAB2044006

  SDS
alfa aesar™
B2044006
5g Each for $145.70

AAB2044014

  SDS
alfa aesar™
B2044014
25g Each for $580.11
Description & Specifications

Specifications

Formula Weight 116.16
Chemical Name or Material 3-Methyl-1,2-cyclopentanediol
Name Note mixture of diastereomers
Assay 95%
Molecular Formula C6H12O2
MDL Number MFCD00019285
CAS 27583-37-5
Quantity 1g