missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Methoxythiophene, TCI America™

Manufacturer:  TCI America M10935G

 View more versions of this product

Catalog No. M10935G



Specifications

Specifications

3-Methoxythiophene
Yellow
17573-92-1
MFCD00043894
thiophene, 3-methoxy, 3-methoxy-thiophene, pubchem8863, acmc-1bow6, 3-methoxythiophene, ksc491i4p, bidd:gt0411
COC1=CSC=C1
114.162
114.16
≥98.0% (GC)
82°C
5g
C5H6OS
1993
RFSKGCVUDQRZSD-UHFFFAOYSA-N
3-methoxythiophene
140249
Clear Liquid at 20°C

Chemical Identifiers

17573-92-1
114.162
RFSKGCVUDQRZSD-UHFFFAOYSA-N
140249
COC1=CSC=C1
C5H6OS
MFCD00043894
thiophene, 3-methoxy, 3-methoxy-thiophene, pubchem8863, acmc-1bow6, 3-methoxythiophene, ksc491i4p, bidd:gt0411
3-methoxythiophene
Safety and Handling

Safety and Handling

EINECSNumber : (5)-6392

TSCA : No

Recommended Storage : Refrigerator

Documents