missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-methoxythiophene-2-carbaldehyde, 97%, Maybridge™

Manufacturer:  Fisher Scientific Company CC68604DA

 View more versions of this product

Catalog No. CC68604DA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Specifications

3-Methoxythiophene-2-carbaldehyde
1g
96.5
97%
MFCD01571098
KGJDTMQUUPIAEF-UHFFFAOYSA-N
3-methoxythiophene-2-carbaldehyde
588315
97%
Amber Glass Bottle
35134-07-7
100.0
C6H6O2S
2-formyl-3-methoxythiophene, 2-thiophenecarboxaldehyde, 3-methoxy, 2-thiophenecarboxaldehyde,3-methoxy, 3-methoxy-2-thenaldehyde, 3-methoxy-2-thiophenealdehyde, 3-methoxy-2-thiophenecarbaldehyde, 3-methoxy-2-thiophenecarbaldehyde #, 3-methoxy-2-thiophenecarboxaldehyde, 3-methoxy-thiophene-2-carbaldehyde, 3-methoxythiophene-2-carboxaldehyde
COC1=C(SC=C1)C=O
142.172
142.18

Chemical Identifiers

35134-07-7
142.172
KGJDTMQUUPIAEF-UHFFFAOYSA-N
588315
COC1=C(SC=C1)C=O
C6H6O2S
MFCD01571098
2-formyl-3-methoxythiophene, 2-thiophenecarboxaldehyde, 3-methoxy, 2-thiophenecarboxaldehyde,3-methoxy, 3-methoxy-2-thenaldehyde, 3-methoxy-2-thiophenealdehyde, 3-methoxy-2-thiophenecarbaldehyde, 3-methoxy-2-thiophenecarbaldehyde #, 3-methoxy-2-thiophenecarboxaldehyde, 3-methoxy-thiophene-2-carbaldehyde, 3-methoxythiophene-2-carboxaldehyde
3-methoxythiophene-2-carbaldehyde
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if swallowed.
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

SDS Documents