missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Methoxythioanisole, TCI America™

$36.67 - $148.20

Chemical Identifiers

CAS 2388-74-1
Molecular Formula C8H10OS
Molecular Weight (g/mol) 154.227
MDL Number MFCD03789162
InChI Key IWVQVOXDIOKVBE-UHFFFAOYSA-N
Synonym 3-(Methylthio)anisole
PubChem CID 281182
IUPAC Name 1-methoxy-3-methylsulfanylbenzene
SMILES COC1=CC(=CC=C1)SC
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M188025G View Documents TCI America
M188025G
25g Each for $148.20
 
M18805G View Documents TCI America
M18805G
5g Each for $36.67
 
Specifications

Chemical Identifiers

2388-74-1
154.227
IWVQVOXDIOKVBE-UHFFFAOYSA-N
281182
COC1=CC(=CC=C1)SC
C8H10OS
MFCD03789162
3-(Methylthio)anisole
1-methoxy-3-methylsulfanylbenzene

Specifications

127°C
25g
C8H10OS
3-(Methylthio)anisole
COC1=CC(=CC=C1)SC
154.227
154.23
≥98.0% (GC)
Yellow
2388-74-1
MFCD03789162
IWVQVOXDIOKVBE-UHFFFAOYSA-N
1-methoxy-3-methylsulfanylbenzene
281182
Clear Liquid at 20°C
3-Methoxythioanisole
Safety and Handling

Safety and Handling

TSCA : No