missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(3-methoxythien-2-yl)methylamine, 95%, Maybridge™

Manufacturer:  Fisher Scientific Company CC68613DA

 View more versions of this product

Catalog No. CC68613DA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

(3-Methoxythien-2-yl)methylamine
1g
94.5
95%
MFCD11841071
GDDVAKBOBBYCLK-UHFFFAOYSA-N
(3-methoxythiophen-2-yl)methanamine
43811049
95%
Amber Glass Bottle
946409-37-6
100.0
C6H9NOS
1-3-methoxythiophen-2-yl methanamine, 2-aminomethyl-3-methoxythiophene, 3-methoxy-2-thienyl methyl amine, 3-methoxy-2-thienyl methylamine, 3-methoxy-2-thiophenyl methanamine, 3-methoxythien-2-yl methylamine, 3-methoxythiophen-2-yl methanamine
COC1=C(SC=C1)CN
143.204
143.21

Chemical Identifiers

946409-37-6
143.204
GDDVAKBOBBYCLK-UHFFFAOYSA-N
43811049
COC1=C(SC=C1)CN
C6H9NOS
MFCD11841071
1-3-methoxythiophen-2-yl methanamine, 2-aminomethyl-3-methoxythiophene, 3-methoxy-2-thienyl methyl amine, 3-methoxy-2-thienyl methylamine, 3-methoxy-2-thiophenyl methanamine, 3-methoxythien-2-yl methylamine, 3-methoxythiophen-2-yl methanamine
(3-methoxythiophen-2-yl)methanamine
Safety and Handling

Safety and Handling

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

SDS Documents