missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Methoxypyridine, TCI America™

$27.50 - $82.50

Chemical Identifiers

CAS 7295-76-3
Molecular Formula C6H7NO
Molecular Weight (g/mol) 109.128
MDL Number MFCD00673022
InChI Key UMJSCPRVCHMLSP-UHFFFAOYSA-N
Synonym pyridine, 3-methoxy, beta-methoxypyridine, 3-methyoxypyridine, unii-xv2a2d8595, .beta.-methoxypyridine, 3-methoxy pyridine, 3-methoxypiridine, 3-methoxy-pyridine, pubchem15370, acmc-209oov
PubChem CID 23719
IUPAC Name 3-methoxypyridine
SMILES COC1=CN=CC=C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M23041G View Documents TCI America
M23041G
1g Each for $27.50
 
M23045G View Documents TCI America
M23045G
5g Each for $82.50
 
Specifications

Chemical Identifiers

7295-76-3
109.128
UMJSCPRVCHMLSP-UHFFFAOYSA-N
23719
COC1=CN=CC=C1
C6H7NO
MFCD00673022
pyridine, 3-methoxy, beta-methoxypyridine, 3-methyoxypyridine, unii-xv2a2d8595, .beta.-methoxypyridine, 3-methoxy pyridine, 3-methoxypiridine, 3-methoxy-pyridine, pubchem15370, acmc-209oov
3-methoxypyridine

Specifications

179°C
1g
C6H7NO
pyridine, 3-methoxy, beta-methoxypyridine, 3-methyoxypyridine, unii-xv2a2d8595, .beta.-methoxypyridine, 3-methoxy pyridine, 3-methoxypiridine, 3-methoxy-pyridine, pubchem15370, acmc-209oov
COC1=CN=CC=C1
109.128
109.13
≥96.0% (GC,T)
Yellow
7295-76-3
MFCD00673022
UMJSCPRVCHMLSP-UHFFFAOYSA-N
3-methoxypyridine
23719
Clear Liquid at 20°C
3-Methoxypyridine
Safety and Handling

Safety and Handling

TSCA : No