missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Methoxypyridazine 97.0+%, TCI America™

$205.73 - $681.39

Chemical Identifiers

CAS 19064-65-4
Molecular Formula C5H6N2O
Molecular Weight (g/mol) 110.116
MDL Number MFCD00234098
InChI Key ASFHDLDAWYTMJS-UHFFFAOYSA-N
PubChem CID 292493
IUPAC Name 3-methoxypyridazine
SMILES COC1=NN=CC=C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M20781G View Documents Tci America
M20781G
1g Each for $205.73
 
M20785G View Documents Tci America
M20785G
5g Each for $681.39
 
Specifications

Chemical Identifiers

19064-65-4
110.116
ASFHDLDAWYTMJS-UHFFFAOYSA-N
3-methoxypyridazine
C5H6N2O
MFCD00234098
292493
COC1=NN=CC=C1

Specifications

87°C
1g
C5H6N2O
ASFHDLDAWYTMJS-UHFFFAOYSA-N
3-methoxypyridazine
292493
Clear Liquid at 20°C
3-Methoxypyridazine
Undesignated
19064-65-4
MFCD00234098
COC1=NN=CC=C1
110.116
110.12
≥97.0% (GC,T)
Safety and Handling

Safety and Handling

TSCA : No