Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Methoxypropylamine, 99%

Chemical Identifiers

Chemical Name or Material 3-Methoxypropylamine
CAS 5332-73-0
Molecular Formula CH3O(CH2)3NH2
Formula Weight 89.14
MDL Number MFCD00014831
UN Number UN2734
Beilstein 878144
Synonym 1-Amino-3-methoxypropane
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB24095AK

 
alfa aesar™
B24095AK
250mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 89.14
Chemical Name or Material 3-Methoxypropylamine
TSCA TSCA
Assay 99%
Molecular Formula CH3O(CH2)3NH2
MDL Number MFCD00014831
Boiling Point 117° to 119°C
CAS 5332-73-0
UN Number UN2734
Beilstein 878144
Melting Point <−70°C
Synonym 1-Amino-3-methoxypropane
Quantity 250mL
Flash Point 27°C (81°F)
Odor Amine-like
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301-H314-H318-H226-H317
Toxic if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapour.
May cause an allergic skin reaction.
Health Hazard 3 GHS P Statement
P210-P301+P310-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, FLAMMABLE, N.O.S.
RTECS Number UI3335000
Safety and Handling

Danger

GHS P Statement
P210-P301+P310-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H314-H318-H226-H317
Toxic if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapour.
May cause an allergic skin reaction.