missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Methoxypropyl isothiocyanate, 98%

Manufacturer:  Alfa Aesar™ A1521614

 View more versions of this product

Catalog No. AAA1521614



Specifications

Specifications

3-Methoxypropyl isothiocyanate
Pungent
25g
C5H9NOS
UN2922
3-methoxypropyl isothiocyanate, 3-methoxypropylisothiocyanate, 1-isothiocyanato-3-methoxy-propane, propane,1-isothiocyanato-3-methoxy, propane, 1-isothiocyanato-3-methoxy, 3-methoxypropanisothiocyanate, acmc-1bu13, 3-methoxy-n-propyl isothiocyanate, 3-methoxpropyl isothiocyanate, #
COCCCN=C=S
131.193
131.2
77°C (170°F)
119°C to 120°C (35mmHg)
17702-11-3
MFCD00022063
1751281
UKVFWFDXYBLOLU-UHFFFAOYSA-N
1-isothiocyanato-3-methoxypropane
140274
98%

Chemical Identifiers

17702-11-3
131.193
UKVFWFDXYBLOLU-UHFFFAOYSA-N
140274
COCCCN=C=S
C5H9NOS
MFCD00022063
3-methoxypropyl isothiocyanate, 3-methoxypropylisothiocyanate, 1-isothiocyanato-3-methoxy-propane, propane,1-isothiocyanato-3-methoxy, propane, 1-isothiocyanato-3-methoxy, 3-methoxypropanisothiocyanate, acmc-1bu13, 3-methoxy-n-propyl isothiocyanate, 3-methoxpropyl isothiocyanate, #
1-isothiocyanato-3-methoxypropane
Safety and Handling

Safety and Handling

GHS H Statement
H331-H314-H318-H227-H302-H312
Toxic if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
Harmful if swallowed.
Harmful in contact with skin.

GHS P Statement
P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUIDS, TOXIC, N.O.S.

Documents