Learn More
Alfa Aesar™ 3-Methoxypropyl isothiocyanate, 98%
Manufacturer: Alfa Aesar™ A1521606
Specifications
3-Methoxypropyl isothiocyanate | |
Pungent | |
5g | |
C5H9NOS | |
UN2922 | |
3-methoxypropyl isothiocyanate, 3-methoxypropylisothiocyanate, 1-isothiocyanato-3-methoxy-propane, propane,1-isothiocyanato-3-methoxy, propane, 1-isothiocyanato-3-methoxy, 3-methoxypropanisothiocyanate, acmc-1bu13, 3-methoxy-n-propyl isothiocyanate, 3-methoxpropyl isothiocyanate, # | |
COCCCN=C=S | |
131.193 | |
131.2 |
77°C (170°F) | |
119°C to 120°C (35mmHg) | |
17702-11-3 | |
MFCD00022063 | |
1751281 | |
UKVFWFDXYBLOLU-UHFFFAOYSA-N | |
1-isothiocyanato-3-methoxypropane | |
140274 | |
98% |
Chemical Identifiers
17702-11-3 | |
131.193 | |
UKVFWFDXYBLOLU-UHFFFAOYSA-N | |
140274 | |
COCCCN=C=S |
C5H9NOS | |
MFCD00022063 | |
3-methoxypropyl isothiocyanate, 3-methoxypropylisothiocyanate, 1-isothiocyanato-3-methoxy-propane, propane,1-isothiocyanato-3-methoxy, propane, 1-isothiocyanato-3-methoxy, 3-methoxypropanisothiocyanate, acmc-1bu13, 3-methoxy-n-propyl isothiocyanate, 3-methoxpropyl isothiocyanate, # | |
1-isothiocyanato-3-methoxypropane |
Safety and Handling
GHS H Statement
H331-H314-H318-H227-H302-H312
Toxic if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
Harmful if swallowed.
Harmful in contact with skin.
GHS P Statement
P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Danger
DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUIDS, TOXIC, N.O.S.