Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Methoxypropyl isothiocyanate, 98%

Chemical Identifiers

Chemical Name or Material 3-Methoxypropyl isothiocyanate
CAS 17702-11-3
Molecular Formula CH3O(CH2)3NCS
Formula Weight 131.2
MDL Number MFCD00022063
UN Number UN2922
Beilstein 1751281
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1521606

  SDS
alfa aesar™
A1521606
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1521614

  SDS
alfa aesar™
A1521614
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 131.2
Chemical Name or Material 3-Methoxypropyl isothiocyanate
Assay 98%
Molecular Formula CH3O(CH2)3NCS
MDL Number MFCD00022063
Boiling Point 119° to 120°C (35mmHg)
CAS 17702-11-3
UN Number UN2922
Beilstein 1751281
Quantity 5g
Flash Point 77°C (170°F)
Odor Pungent
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H331-H314-H318-H227-H302-H312
Toxic if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
Harmful if swallowed.
Harmful in contact with skin.
Health Hazard 3 GHS P Statement
P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUIDS, TOXIC, N.O.S.
Safety and Handling

Danger

GHS P Statement
P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H331-H314-H318-H227-H302-H312
Toxic if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
Harmful if swallowed.
Harmful in contact with skin.