Please login to your online account to display your discounted pricing

3-Methoxypropyl isothiocyanate, 97%, Maybridge


Quantity 10g
Packaging Amber glass bottle

Manufacturer: fisher scientific company  TL04220EA

 View more versions of this product

Catalog No. TL04220EA

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 131.2
Quantity 10g
Assay 97%
CAS Min % 97.0
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Chemical Name or Material 3-Methoxypropyl isothiocyanate
Molecular Formula C5H9NOS
CAS 17702-11-3
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Packaging Amber glass bottle
Assay Percent Range 97%
MDL Number MFCD00022063

Specifications

Chemical Name or Material 3-Methoxypropyl isothiocyanate
CAS 17702-11-3
CAS Min % 97.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C5H9NOS
Formula Weight 131.2
MDL Number MFCD00022063
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.