missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
3-Methoxypropionitrile, 98%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific 257441000
Specifications
3-Methoxypropionitrile | |
66°C | |
Glass bottle | |
0.938 | |
Colorless to Yellow | |
100mL | |
110-67-8 | |
100.0 | |
C4H7NO | |
MFCD00001958 | |
Solubility in water: 335g/L (20°C) | |
COCCC#N | |
85.11 | |
85.11 | |
98% |
0.9380g/mL | |
Authentic | |
1.4020 to 1.4040 | |
164.0°C to 165.0°C | |
-62.9°C | |
98% | |
97.5 | |
98% | |
CH3OCH2CH2CN | |
1-cyano-2-methoxyethane, 2-cyanoethyl methyl ether, 3-methoxypropannitril, 3-methoxypropionitrile, 3-methoxypropiononitrile, 3-methoxypropylnitrile, beta-methoxypropionitrile, beta-methyoxypropionitrile, propanenitrile, 3-methoxy, propionitrile, 3-methoxy | |
OOWFYDWAMOKVSF-UHFFFAOYSA-N | |
3-methoxypropanenitrile | |
61032 | |
Liquid |
Chemical Identifiers
110-67-8 | |
85.11 | |
OOWFYDWAMOKVSF-UHFFFAOYSA-N | |
61032 | |
COCCC#N |
C4H7NO | |
MFCD00001958 | |
1-cyano-2-methoxyethane, 2-cyanoethyl methyl ether, 3-methoxypropannitril, 3-methoxypropionitrile, 3-methoxypropiononitrile, 3-methoxypropylnitrile, beta-methoxypropionitrile, beta-methyoxypropionitrile, propanenitrile, 3-methoxy, propionitrile, 3-methoxy | |
3-methoxypropanenitrile |
Safety and Handling
GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Wear eye protection/face protection.
Wash face,hands and any exposed skin thoroughly after handling.
If eye irritation persists: Get medical advice/attention.
GHS Signal Word: Warning
EINECSNumber : 203-790-7
RTECSNumber : TZ4870000
TSCA : TSCA