missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Methoxypropionitrile, 98%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 257445000

 View more versions of this product

Catalog No. AC257445000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

3-Methoxypropionitrile
66°C
Glass bottle
0.938
Colorless to Yellow
500mL
110-67-8
100.0
C4H7NO
MFCD00001958
Solubility in water: 335g/L (20°C)
COCCC#N
85.11
85.11
98%
0.9380g/mL
Authentic
1.4020 to 1.4040
164.0°C to 165.0°C
-62.9°C
98%
97.5
98%
CH3OCH2CH2CN
1-cyano-2-methoxyethane, 2-cyanoethyl methyl ether, 3-methoxypropannitril, 3-methoxypropionitrile, 3-methoxypropiononitrile, 3-methoxypropylnitrile, beta-methoxypropionitrile, beta-methyoxypropionitrile, propanenitrile, 3-methoxy, propionitrile, 3-methoxy
OOWFYDWAMOKVSF-UHFFFAOYSA-N
3-methoxypropanenitrile
61032
Liquid

Chemical Identifiers

110-67-8
85.11
OOWFYDWAMOKVSF-UHFFFAOYSA-N
61032
COCCC#N
C4H7NO
MFCD00001958
1-cyano-2-methoxyethane, 2-cyanoethyl methyl ether, 3-methoxypropannitril, 3-methoxypropionitrile, 3-methoxypropiononitrile, 3-methoxypropylnitrile, beta-methoxypropionitrile, beta-methyoxypropionitrile, propanenitrile, 3-methoxy, propionitrile, 3-methoxy
3-methoxypropanenitrile
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear eye protection/face protection.
Wash face,hands and any exposed skin thoroughly after handling.
If eye irritation persists: Get medical advice/attention.

GHS Signal Word: Warning

EINECSNumber : 203-790-7

RTECSNumber : TZ4870000

TSCA : TSCA

SDS Documents