missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Methoxypiperidine hydrochloride

$71.72 - $257.05

Chemical Identifiers

CAS 688809-94-1
Molecular Formula C6H13NO·HCl
Formula Weight 151.64
MDL Number MFCD06800959
Chemical Name or Material 3-Methoxypiperidine hydrochloride
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH5113803 SDS Alfa Aesar™
H5113803
1g Each for $257.05
AAH51138MD SDS Alfa Aesar™
H51138MD
250mg Each for $71.72
Specifications Description & Specifications

Chemical Identifiers

688809-94-1
151.64
3-Methoxypiperidine hydrochloride
C6H13NO·HCl
MFCD06800959

Specifications

688809-94-1
151.64
1g
C6H13NO·HCl
MFCD06800959
3-Methoxypiperidine hydrochloride

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning