Please login to your online account to display your discounted pricing

3-Methoxyphenylmagnesium bromide, 1M solution in THF/toluene, ACROS Organics™


Quantity 800mL
Packaging AcroSeal™ Glass Bottle

Manufacturer: thermo fisher scientific  AC426758000

 View more versions of this product

Catalog No. AC426758000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Solution
Formula Weight 211.34
Quantity 800mL
Color Clear yellow-brown
Packaging AcroSeal™ Glass Bottle
Density 1.0000g/mL
UN Number 2924
Additional Information magnesium salts may crystallize below 20°C
Chemical Name or Material 3-Methoxyphenylmagnesium bromide
Name Note 1M solution in THF/toluene
Molecular Formula C7H7BrMgO
Linear Formula CH3OC6H4MgBr
CAS 36282-40-3
CAS Min % 19.02
Health Hazard 2 GHS P Statement May cause respiratory irritation. May cause drowsiness or dizziness. Causes severe skin burns and eye damage. Suspected of damaging fertility or the unborn child. May be fatal if swallowed and enters a
CAS 108-88-3
MDL Number MFCD00672002
CAS Max % 23.25
CAS 109-99-9
CAS Min % 76.75
CAS Max % 80.98
Health Hazard 1 Danger
Health Hazard 3 GHS P Statement Wear eye protection/face protection. IF SWALLOWED: rinse mouth. Do NOT induce vomiting. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Co

Specifications

Chemical Name or Material 3-Methoxyphenylmagnesium bromide
Name Note 1M solution in THF/toluene
CAS 36282-40-3
CAS Min % 19.02
CAS Max % 23.25
CAS 109-99-9
CAS Min % 76.75
CAS Max % 80.98
CAS 108-88-3
Molecular Formula C7H7BrMgO
Linear Formula CH3OC6H4MgBr
Formula Weight 211.34
MDL Number MFCD00672002
Accessories

Danger

GHS P Statement Wear eye protection/face protection. IF SWALLOWED: rinse mouth. Do NOT induce vomiting. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Co

GHS P Statement May cause respiratory irritation. May cause drowsiness or dizziness. Causes severe skin burns and eye damage. Suspected of damaging fertility or the unborn child. May be fatal if swallowed and enters a

SDS & Safety Documents