missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Methoxyphenylmagnesium bromide, 1M solution in THF/toluene, AcroSeal™, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 426751000

 View more versions of this product

Catalog No. AC426751000


Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

3-Methoxyphenylmagnesium bromide
36282-40-3,109-99-9,108-88-3
23.25,80.98
1.0000g/mL
AcroSeal™ Glass bottle
CH3OC6H4MgBr
100 mL
1
AJKNYEHIJAMGHX-UHFFFAOYSA-M
magnesium;methoxybenzene;bromide
4605969
Solution
1M solution in THF/toluene
19.02,76.75
Yellow to Brown
−21°C
C7H7BrMgO
MFCD00672002
2924
3-methoxyphenylmagnesium bromide, grignard reagent, 3-methoxyphenylmagnesium bromide solution, 3-methoxyphenylmagnesium bromide solution, 3-methoxyphenylmagnesiumbromid, 3-methoxyphenylmagnesiumbromid, bromo 3-methoxyphenyl magnesium, 3-methoxy phenylmagnesium bromide, m-methoxyphenyl magnesium bromide, m-methoxyphenyl magnesium bromide, 3-methoxylphenyl magnesium bromide
COC1=CC=CC([Mg]Br)=C1
211.34
211.34

Chemical Identifiers

36282-40-3
211.34
AJKNYEHIJAMGHX-UHFFFAOYSA-M
4605969
C7H7BrMgO
MFCD00672002
3-methoxyphenylmagnesium bromide, grignard reagent, 3-methoxyphenylmagnesium bromide solution, 3-methoxyphenylmagnesium bromide solution, 3-methoxyphenylmagnesiumbromid, 3-methoxyphenylmagnesiumbromid, bromo 3-methoxyphenyl magnesium, 3-methoxy phenylmagnesium bromide, m-methoxyphenyl magnesium bromide, m-methoxyphenyl magnesium bromide, 3-methoxylphenyl magnesium bromide
COC1=CC=CC([Mg]Br)=C1
Safety and Handling

Safety and Handling

GHS H Statement
May cause respiratory irritation.
May cause drowsiness or dizziness.
Causes severe skin burns and eye damage.
Suspected of damaging the unborn child.
May be fatal if swallowed and enters airways.
May c

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Wear protective gloves/protective clothing/eye p

GHS Signal Word: Danger

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only