Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Methoxyphenylhydrazine hydrochloride, 98%

$83.18 - $266.70

Chemical Identifiers

Chemical Name or Material 3-Methoxyphenylhydrazine hydrochloride
CAS 39232-91-2
Molecular Formula C7H10N2O·HCl
Formula Weight 174.63
MDL Number MFCD00044416
Beilstein 5304389
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0809103

  SDS
alfa aesar™
L0809103
1g Each for $83.18

AAL0809106

  SDS
alfa aesar™
L0809106
5g Each for $266.70
Description & Specifications

Specifications

Formula Weight 174.63
MDL Number MFCD00044416
Assay 98%
Health Hazard 3 GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Health Hazard 2 GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Chemical Name or Material 3-Methoxyphenylhydrazine hydrochloride
CAS 39232-91-2
Melting Point ≈142°C
Health Hazard 1 Danger
Molecular Formula C7H10N2O·HCl
Recommended Storage −30° to −10°C
Quantity 1g
Beilstein 5304389
Safety and Handling

Danger

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

−30° to −10°C