missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Methoxyphenylacetylene, 96%

Manufacturer:  Alfa Aesar™ H5525403

 View more versions of this product

Catalog No. AAH5525403



Description

Description

Applications
It is a useful aromatic compound.

Solubility
Slightly soluble in water.

Notes
Storage: Keep container tightly closed. Store in a cool, dark and well-ventilated place and away from oxidizing agents.
Specifications

Specifications

3-Methoxyphenylacetylene
204°C to 210°C
768-70-7
MFCD00160810
ZASXCTCNZKFDTP-UHFFFAOYSA-N
1-ethynyl-3-methoxybenzene
640753
96%
76°C (169°F)
1g
C9H8O
1-ethynyl-3-methoxy-benzene
COC1=CC=CC(=C1)C#C
132.162
132.16

Chemical Identifiers

768-70-7
132.162
ZASXCTCNZKFDTP-UHFFFAOYSA-N
640753
COC1=CC=CC(=C1)C#C
C9H8O
MFCD00160810
1-ethynyl-3-methoxy-benzene
1-ethynyl-3-methoxybenzene
Safety and Handling

Safety and Handling

GHS H Statement
H227
Combustible liquid.

GHS P Statement
P210-P280-P370+P378a-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
In case of fire: Use for extinction: CO2, powder or water spray.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Recommended Storage : 2° to 8°C

Documents