Please login to your online account to display your discounted pricing

3-Methoxyphenylacetyl chloride, 98%, ACROS Organics™

$85.13 - $85.13

Chemical Identifiers

Chemical Name or Material 3-Methoxyphenylacetyl chloride
CAS 6834-42-0
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C9H9ClO2
Linear Formula CH3OC6H4CHCOCl
Formula Weight 184.62
MDL Number MFCD00012244
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC305910050

  SDS
thermo fisher scientific
305910050
Glass bottle 5g Each for $85.13
Description & Specifications

Specifications

Assay ≥97.5%
Formula Weight 184.62
Physical Form Liquid
Quantity 5g
Color Clear yellow to red-brown
Density 1.18g/mL
CAS Max % 100.0
Packaging Glass bottle
Infrared Spectrum Authentic
Linear Formula CH3OC6H4CHCOCl
CAS 6834-42-0
Chemical Name or Material 3-Methoxyphenylacetyl chloride
Molecular Formula C9H9ClO2
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
MDL Number MFCD00012244
CAS Min % 97.5
Refractive Index 1.5382 to 1.5402 (20°C, 589nm)
Assay Percent Range 98%
Health Hazard 2 GHS H Statement:
Causes severe skin burns and eye damage.
May cause respiratory irritation.
Reacts violently with water.