Please login to your online account to display your discounted pricing

3-Methoxyphenylacetyl chloride, 98%, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  305910050

 View more versions of this product

Catalog No. AC305910050

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥97.5%
Physical Form Liquid
Formula Weight 184.62
Quantity 5g
Color Clear yellow to red-brown
Density 1.18g/mL
CAS Max % 100.0
Packaging Glass bottle
Chemical Name or Material 3-Methoxyphenylacetyl chloride
Infrared Spectrum Authentic
Molecular Formula C9H9ClO2
Linear Formula CH3OC6H4CHCOCl
CAS 6834-42-0
MDL Number MFCD00012244
CAS Min % 97.5
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Refractive Index 1.5382 to 1.5402 (20°C, 589nm)
Assay Percent Range 98%
Health Hazard 2 GHS H Statement:
Causes severe skin burns and eye damage.
May cause respiratory irritation.
Reacts violently with water.

Specifications

Chemical Name or Material 3-Methoxyphenylacetyl chloride
CAS 6834-42-0
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C9H9ClO2
Linear Formula CH3OC6H4CHCOCl
Formula Weight 184.62
MDL Number MFCD00012244
Accessories

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes severe skin burns and eye damage.
May cause respiratory irritation.
Reacts violently with water.

SDS & Safety Documents