missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
3-Methoxyphenylacetic acid, 99.5%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific 126010250
Specifications
3-Methoxyphenylacetic acid | |
Glass bottle | |
71.0°C to 73.0°C | |
1798-09-0 | |
100.0 | |
C9H10O3 | |
MFCD00004334 | |
2-3-methoxyphenyl acetic acid, 25xlo0t6my, 3-methoxy-phenyl-acetic acid, 3-methoxybenzeneacetic acid, 3-methoxyphenyl acetic acid, 3-methoxyphenylacetic acid, acetic acid, m-methoxyphenyl, benzeneacetic acid, 3-methoxy, m-methoxyphenylacetic acid, unii-25xlo0t6my | |
LEGPZHPSIPPYIO-UHFFFAOYSA-N | |
2-(3-methoxyphenyl)acetic acid | |
15719 | |
Flakes |
Authentic | |
White to Yellow | |
25g | |
99.4 | |
99.50% | |
CH3OC6H4CH2CO2H | |
10, I, 82 | |
Solubility in water: slightly soluble. Other solubilities: soluble in ethanol | |
COC1=CC=CC(=C1)CC(=O)O | |
166.18 | |
166.18 | |
99.5% |
Chemical Identifiers
1798-09-0 | |
166.18 | |
LEGPZHPSIPPYIO-UHFFFAOYSA-N | |
15719 | |
COC1=CC=CC(=C1)CC(=O)O |
C9H10O3 | |
MFCD00004334 | |
2-3-methoxyphenyl acetic acid, 25xlo0t6my, 3-methoxy-phenyl-acetic acid, 3-methoxybenzeneacetic acid, 3-methoxyphenyl acetic acid, 3-methoxyphenylacetic acid, acetic acid, m-methoxyphenyl, benzeneacetic acid, 3-methoxy, m-methoxyphenylacetic acid, unii-25xlo0t6my | |
2-(3-methoxyphenyl)acetic acid |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
If eye irritation persists: Get medical advice/attention.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER
GHS Signal Word: Warning
EINECSNumber : 217-282-8