missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Methoxyphenylacetic acid, 97%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A1095106

 View more versions of this product

Catalog No. AAA1095106



Description

Description

A fluorimetric method for the estimation of 4-hydroxy-3-methoxyphenylacetic acid (homovanillic acid) has been developed and applied to normal brain tissue. The presence of homovanillic acid in the caudate nucleus of normal animals of several species has been demonstrated.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
A fluorimetric method for the estimation of 4-hydroxy-3-methoxyphenylacetic acid (homovanillic acid) has been developed and applied to normal brain tissue. The presence of homovanillic acid in the caudate nucleus of normal animals of several species has been demonstrated.

Solubility
Soluble in Chloroform and Ethyl Acetate.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Specifications

Specifications

3-Methoxyphenylacetic acid
67°C to 72°C
MFCD00004334
2614004
Soluble in Chloroform and Ethyl Acetate.
COC1=CC=CC(=C1)CC(=O)O
166.176
166.18
1798-09-0
C9H10O3
5 g
3-methoxyphenylacetic acid, 2-3-methoxyphenyl acetic acid, m-methoxyphenylacetic acid, 3-methoxyphenyl acetic acid, 3-methoxybenzeneacetic acid, benzeneacetic acid, 3-methoxy, acetic acid, m-methoxyphenyl, unii-25xlo0t6my, 3-methoxy-phenyl-acetic acid, 25xlo0t6my
LEGPZHPSIPPYIO-UHFFFAOYSA-N
2-(3-methoxyphenyl)acetic acid
15719
97%

Chemical Identifiers

1798-09-0
166.176
LEGPZHPSIPPYIO-UHFFFAOYSA-N
15719
COC1=CC=CC(=C1)CC(=O)O
C9H10O3
MFCD00004334
3-methoxyphenylacetic acid, 2-3-methoxyphenyl acetic acid, m-methoxyphenylacetic acid, 3-methoxyphenyl acetic acid, 3-methoxybenzeneacetic acid, benzeneacetic acid, 3-methoxy, acetic acid, m-methoxyphenyl, unii-25xlo0t6my, 3-methoxy-phenyl-acetic acid, 25xlo0t6my
2-(3-methoxyphenyl)acetic acid
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 217-282-8

RTECSNumber : AI8940000

TSCA : No

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only