Please login to your online account to display your discounted pricing

3-Methoxyphenyl isocyanate, 99%, ACROS Organics™


Quantity 100g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  221161000

 View more versions of this product

Catalog No. AC221161000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥98.5%
Physical Form Liquid
Formula Weight 149.15
Quantity 100g
Color Clear colorless to slightly pink
Beilstein 13, III, 963
EINECS Number 242-658-3
TSCA TSCA
Density 1.13g/mL
CAS Max % 100.0
Chemical Name or Material 3-Methoxyphenyl isocyanate
Infrared Spectrum Authentic
Packaging Glass bottle
Name Note 99%
Assay Percent Range 99%
Molecular Formula C8H7NO2
Linear Formula CH3OC6H4NCO
MDL Number MFCD00002019
CAS 18908-07-1
CAS Min % 98.5
Health Hazard 1 GHS Signal Word: Warning
Health Hazard 2 GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Refractive Index 1.5420 to 1.5440 (20°C, 589nm)

Specifications

Chemical Name or Material 3-Methoxyphenyl isocyanate
Name Note 99%
CAS 18908-07-1
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C8H7NO2
Linear Formula CH3OC6H4NCO
Formula Weight 149.15
MDL Number MFCD00002019
Accessories

GHS Signal Word: Warning

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

SDS & Safety Documents