Please login to your online account to display your discounted pricing

(3-Methoxyphenyl)acetonitrile, 97%, ACROS Organics™


Quantity 50g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC197870500

 View more versions of this product

Catalog No. AC197870500

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥96.0%
Physical Form Liquid
Formula Weight 147.18
Quantity 50g
Color Clear colorless to slightly yellow
EINECS Number 243-428-5
TSCA TSCA
Density 1.0500g/mL
Beilstein 10, III, 429
CAS Min % 96.0
CAS Max % 100.0
Chemical Name or Material (3-Methoxyphenyl)acetonitrile
Infrared Spectrum Authentic
Packaging Glass bottle
Synonym m-Methoxybenzyl cyanide
Health Hazard 1 GHS Signal Word: Warning
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Refractive Index 1.5297 to 1.5317 (20°C, 589nm)
Name Note 97%
Assay Percent Range 97%
Molecular Formula C9H9NO
Linear Formula CH3OC6H4CH2CN
MDL Number MFCD00001910
CAS 19924-43-7

Specifications

Chemical Name or Material (3-Methoxyphenyl)acetonitrile
Name Note 97%
CAS 19924-43-7
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C9H9NO
Linear Formula CH3OC6H4CH2CN
Formula Weight 147.18
MDL Number MFCD00001910
Synonym m-Methoxybenzyl cyanide
Accessories

GHS Signal Word: Warning

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.

SDS & Safety Documents