missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Methoxyphenol, TCI America™

Manufacturer:  TCI America M0122500ML

 View more versions of this product

Catalog No. M0122500ML



Specifications

Specifications

3-Methoxyphenol
Yellow
150-19-6
MFCD00002267
m-methoxyphenol, 3-hydroxyanisole, m-hydroxyanisole, phenol, 3-methoxy, m-guaiacol, resorcinol monomethyl ether, resorcinol methyl ether, 1-hydroxy-3-methoxybenzene, phenol, m-methoxy, 3-methoxy phenol
COC1=CC=CC(=C1)O
124.139
CHEBI:52678
Clear Liquid at 20°C
244°C
500mL
C7H8O2
2810
ASHGTJPOSUFTGB-UHFFFAOYSA-N
3-methoxyphenol
9007
124.14
≥98.0% (GC)

Chemical Identifiers

150-19-6
124.139
ASHGTJPOSUFTGB-UHFFFAOYSA-N
9007
3-methoxyphenol
C7H8O2
MFCD00002267
m-methoxyphenol, 3-hydroxyanisole, m-hydroxyanisole, phenol, 3-methoxy, m-guaiacol, resorcinol monomethyl ether, resorcinol methyl ether, 1-hydroxy-3-methoxybenzene, phenol, m-methoxy, 3-methoxy phenol
CHEBI:52678
COC1=CC=CC(=C1)O
Safety and Handling

Safety and Handling

EINECSNumber : (3)-0567

RTECSNumber : SL7524000

TSCA : Yes

Documents