Learn More
3-Methoxyphenol, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 161065000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
3-Methoxyphenol | |
96.0 | |
-17.0°C | |
1.1450g/mL | |
>110°C | |
96% min. (GC) | |
C7H8O2 | |
CH3OC6H4OH | |
500 mL | |
13,181 | |
m-methoxyphenol, 3-hydroxyanisole, m-hydroxyanisole, phenol, 3-methoxy, m-guaiacol, resorcinol monomethyl ether, resorcinol methyl ether, 1-hydroxy-3-methoxybenzene, phenol, m-methoxy, 3-methoxy phenol | |
ASHGTJPOSUFTGB-UHFFFAOYSA-N | |
3-methoxyphenol | |
9007 | |
124.14 | |
Liquid |
150-19-6 | |
100.0 | |
Brown to Pink-Red | |
244.0°C | |
Authentic | |
Glass bottle | |
1.5510 to 1.5530 | |
MFCD00002267 | |
06, 813 | |
1.145 | |
Solubility in water: slightly soluble | |
COC1=CC=CC(=C1)O | |
124.14 | |
CHEBI:52678 | |
97% |
Chemical Identifiers
150-19-6 | |
124.14 | |
ASHGTJPOSUFTGB-UHFFFAOYSA-N | |
9007 | |
3-methoxyphenol |
C7H8O2 | |
MFCD00002267 | |
m-methoxyphenol, 3-hydroxyanisole, m-hydroxyanisole, phenol, 3-methoxy, m-guaiacol, resorcinol monomethyl ether, resorcinol methyl ether, 1-hydroxy-3-methoxybenzene, phenol, m-methoxy, 3-methoxy phenol | |
CHEBI:52678 | |
COC1=CC=CC(=C1)O |
Safety and Handling
GHS H Statement
Causes skin irritation.
Harmful if swallowed.
May cause respiratory irritation.
Toxic in contact with skin.
Causes serious eye damage.
Harmful if inhaled.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Immediately call a POISON CENTER or doc
GHS Signal Word: Danger
EINECSNumber : 205-754-6
RUO – Research Use Only