missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Methoxyphenol, 97%, ACROS Organics™

Manufacturer:  Acros Organics 161060050

 View more versions of this product

Catalog No. AC161060050



Specifications

Specifications

3-Methoxyphenol
>110°C
Glass bottle
244.0°C
-17.0°C
150-19-6
100.0
C7H8O2
MFCD00002267
13,181
ASHGTJPOSUFTGB-UHFFFAOYSA-N
3-methoxyphenol
9007
124.14
97%
1.1450g/mL
Authentic
1.5510 to 1.5530
Brown to Pink-Red
5mL
96.0
97%
CH3OC6H4OH
06, 813
m-methoxyphenol, 3-hydroxyanisole, m-hydroxyanisole, phenol, 3-methoxy, m-guaiacol, resorcinol monomethyl ether, resorcinol methyl ether, 1-hydroxy-3-methoxybenzene, phenol, m-methoxy, 3-methoxy phenol
COC1=CC=CC(=C1)O
124.139
CHEBI:52678
Liquid

Chemical Identifiers

150-19-6
124.139
ASHGTJPOSUFTGB-UHFFFAOYSA-N
9007
3-methoxyphenol
C7H8O2
MFCD00002267
m-methoxyphenol, 3-hydroxyanisole, m-hydroxyanisole, phenol, 3-methoxy, m-guaiacol, resorcinol monomethyl ether, resorcinol methyl ether, 1-hydroxy-3-methoxybenzene, phenol, m-methoxy, 3-methoxy phenol
CHEBI:52678
COC1=CC=CC(=C1)O
Safety and Handling

Safety and Handling

GHS P Statement
Causes skin irritation.
Harmful if swallowed.
May cause respiratory irritation.
Harmful in contact with skin.
Causes serious eye irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning

SDS Documents