missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
3-Methoxymethylindole 98.0+%, TCI America™
$187.46 - $2163.33
Chemical Identifiers
CAS | 78440-76-3 |
---|---|
Molecular Formula | C10H11NO |
Molecular Weight (g/mol) | 161.204 |
MDL Number | MFCD00276420 |
InChI Key | RJWLFBCRNXZXTJ-UHFFFAOYSA-N |
PubChem CID | 154411 |
IUPAC Name | 3-(methoxymethyl)-1H-indole |
SMILES | COCC1=CNC2=CC=CC=C21 |
Chemical Identifiers
78440-76-3 | |
161.204 | |
RJWLFBCRNXZXTJ-UHFFFAOYSA-N | |
3-(methoxymethyl)-1H-indole |
C10H11NO | |
MFCD00276420 | |
154411 | |
COCC1=CNC2=CC=CC=C21 |
Specifications
78440-76-3 | |
C10H11NO | |
1 g | |
COCC1=CNC2=CC=CC=C21 | |
161.204 | |
161.20 | |
Crystalline Powder |
98°C | |
MFCD00276420 | |
RJWLFBCRNXZXTJ-UHFFFAOYSA-N | |
3-(methoxymethyl)-1H-indole | |
154411 | |
≥98.0% (HPLC,N) | |
3-Methoxymethylindole |
Safety and Handling
TSCA : No