missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-(Methoxymethoxy)benzaldehyde, TCI America™

$220.00 - $721.13

Chemical Identifiers

CAS 13709-05-2
Molecular Formula C9H10O3
Molecular Weight (g/mol) 166.176
MDL Number MFCD04038425
InChI Key JAFJNSQISURLCX-UHFFFAOYSA-N
PubChem CID 12273513
IUPAC Name 3-(methoxymethoxy)benzaldehyde
SMILES COCOC1=CC=CC(=C1)C=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M14485G View Documents TCI America
M14485G
5g Each for $220.00
 
M144825G View Documents TCI America
M144825G
25g Each for $721.13
 
Specifications

Chemical Identifiers

13709-05-2
166.176
JAFJNSQISURLCX-UHFFFAOYSA-N
3-(methoxymethoxy)benzaldehyde
C9H10O3
MFCD04038425
12273513
COCOC1=CC=CC(=C1)C=O

Specifications

Yellow
13709-05-2
MFCD04038425
COCOC1=CC=CC(=C1)C=O
166.176
166.18
≥95.0% (GC)
5g
C9H10O3
JAFJNSQISURLCX-UHFFFAOYSA-N
3-(methoxymethoxy)benzaldehyde
12273513
Clear Liquid at 20°C
3-(Methoxymethoxy)benzaldehyde
Safety and Handling

Safety and Handling

TSCA : No