missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Methoxyflavone, TCI America™

$172.64 - $557.66

Chemical Identifiers

CAS 7245-02-5
Molecular Formula C16H12O3
Molecular Weight (g/mol) 252.27
MDL Number MFCD00017612
InChI Key ZAIANDVQAMEDFL-UHFFFAOYSA-N
PubChem CID 146013
IUPAC Name 3-methoxy-2-phenylchromen-4-one
SMILES COC1=C(OC2=CC=CC=C2C1=O)C3=CC=CC=C3
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M23641G View Documents TCI America
M23641G
1g Each for $172.64
 
M23645G View Documents TCI America
M23645G
5g Each for $557.66
 
Specifications

Chemical Identifiers

7245-02-5
252.27
ZAIANDVQAMEDFL-UHFFFAOYSA-N
3-methoxy-2-phenylchromen-4-one
C16H12O3
MFCD00017612
146013
COC1=C(OC2=CC=CC=C2C1=O)C3=CC=CC=C3

Specifications

White-Yellow
1g
C16H12O3
ZAIANDVQAMEDFL-UHFFFAOYSA-N
3-methoxy-2-phenylchromen-4-one
146013
Crystal-Powder at 20°C
3-Methoxyflavone
114°C
7245-02-5
MFCD00017612
COC1=C(OC2=CC=CC=C2C1=O)C3=CC=CC=C3
252.27
252.27
≥98.0% (GC)
Safety and Handling

Safety and Handling

TSCA : No