missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Methoxycyclohexene, TCI America™

$326.95 - $1,135.16

Chemical Identifiers

CAS 2699-13-0
Molecular Formula C7H12O
Molecular Weight (g/mol) 112.172
InChI Key OKDKFTKUXADLSJ-UHFFFAOYSA-N
PubChem CID 137685
IUPAC Name 3-methoxycyclohexene
SMILES COC1CCCC=C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M24555ML View Documents TCI America
M24555ML
5mL Each for $1,135.16
 
M24551ML View Documents TCI America
M24551ML
1mL Each for $326.95
 
Specifications

Chemical Identifiers

2699-13-0
112.172
137685
COC1CCCC=C1
C7H12O
OKDKFTKUXADLSJ-UHFFFAOYSA-N
3-methoxycyclohexene

Specifications

133°C
5mL
C7H12O
OKDKFTKUXADLSJ-UHFFFAOYSA-N
3-methoxycyclohexene
137685
Clear Liquid at 20°C
3-Methoxycyclohexene
Colorless
2699-13-0
3271
COC1CCCC=C1
112.172
112.17
≥97.0% (GC)
Safety and Handling

Safety and Handling

TSCA : No