missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
3-Methoxycinnamic Acid 99.0+%, TCI America™
$164.24 - $1186.19
Chemical Identifiers
CAS | 6099-04-3 |
---|---|
Molecular Formula | C10H10O3 |
Molecular Weight (g/mol) | 178.19 |
MDL Number | MFCD00004386 |
InChI Key | LZPNXAULYJPXEH-AATRIKPKSA-N |
Synonym | 3-methoxycinnamic acid, 3-3-methoxyphenyl acrylic acid, 2e-3-3-methoxyphenyl acrylic acid, 2e-3-3-methoxyphenyl prop-2-enoic acid, trans-3-methoxycinnamic acid, m-methoxycinnamic acid, trans-3-3-methoxyphenyl acrylic acid, e-3-3-methoxyphenyl acrylic acid, 2-propenoic acid, 3-3-methoxyphenyl-, 2e, trans-m-methoxycinnamic acid |
PubChem CID | 637668 |
IUPAC Name | (E)-3-(3-methoxyphenyl)prop-2-enoic acid |
SMILES | COC1=CC=CC(=C1)C=CC(=O)O |
Chemical Identifiers
6099-04-3 | |
178.19 | |
LZPNXAULYJPXEH-AATRIKPKSA-N | |
637668 | |
COC1=CC=CC(=C1)C=CC(=O)O |
C10H10O3 | |
MFCD00004386 | |
3-methoxycinnamic acid, 3-3-methoxyphenyl acrylic acid, 2e-3-3-methoxyphenyl acrylic acid, 2e-3-3-methoxyphenyl prop-2-enoic acid, trans-3-methoxycinnamic acid, m-methoxycinnamic acid, trans-3-3-methoxyphenyl acrylic acid, e-3-3-methoxyphenyl acrylic acid, 2-propenoic acid, 3-3-methoxyphenyl-, 2e, trans-m-methoxycinnamic acid | |
(E)-3-(3-methoxyphenyl)prop-2-enoic acid |
Specifications
6099-04-3 | |
White | |
MFCD00004386 | |
3-methoxycinnamic acid, 3-3-methoxyphenyl acrylic acid, 2e-3-3-methoxyphenyl acrylic acid, 2e-3-3-methoxyphenyl prop-2-enoic acid, trans-3-methoxycinnamic acid, m-methoxycinnamic acid, trans-3-3-methoxyphenyl acrylic acid, e-3-3-methoxyphenyl acrylic acid, 2-propenoic acid, 3-3-methoxyphenyl-, 2e, trans-m-methoxycinnamic acid | |
COC1=CC=CC(=C1)C=CC(=O)O | |
178.19 | |
178.19 | |
Crystalline Powder |
119°C | |
C10H10O3 | |
25 g | |
LZPNXAULYJPXEH-AATRIKPKSA-N | |
(E)-3-(3-methoxyphenyl)prop-2-enoic acid | |
637668 | |
≥99.0% (GC,T) | |
3-Methoxycinnamic Acid |
Safety and Handling
TSCA : No