missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Methoxycatechol, TCI America™

$174.17 - $1,443.78

Chemical Identifiers

CAS 934-00-9
Molecular Formula C7H8O3
Molecular Weight (g/mol) 140.138
MDL Number MFCD00002191
InChI Key LPYUENQFPVNPHY-UHFFFAOYSA-N
Synonym 3-methoxycatechol, 1,2-benzenediol, 3-methoxy, 3-methoxypyrocatechol, pyrocatechol, 3-methoxy, 2,3-dihydroxyanisole, pyrogallol 1-methyl ether, 1,2-dihydroxy-3-methoxybenzene, 6-methoxycatechol, pyrogallol 1-monomethyl ether, 3-methoxy-1,2-benzenediol
PubChem CID 13622
IUPAC Name 3-methoxybenzene-1,2-diol
SMILES COC1=CC=CC(=C1O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M052425G View Documents TCI America
M052425G
25g Each for $174.17
 
M0524500G View Documents TCI America
M0524500G
500g Each for $1,443.78
 
Specifications

Chemical Identifiers

934-00-9
140.138
LPYUENQFPVNPHY-UHFFFAOYSA-N
13622
COC1=CC=CC(=C1O)O
C7H8O3
MFCD00002191
3-methoxycatechol, 1,2-benzenediol, 3-methoxy, 3-methoxypyrocatechol, pyrocatechol, 3-methoxy, 2,3-dihydroxyanisole, pyrogallol 1-methyl ether, 1,2-dihydroxy-3-methoxybenzene, 6-methoxycatechol, pyrogallol 1-monomethyl ether, 3-methoxy-1,2-benzenediol
3-methoxybenzene-1,2-diol

Specifications

164°C
42°C
934-00-9
MFCD00002191
LPYUENQFPVNPHY-UHFFFAOYSA-N
3-methoxybenzene-1,2-diol
13622
Crystal-Powder at 20°C
3-Methoxycatechol
White-Yellow
25g
C7H8O3
3-methoxycatechol, 1,2-benzenediol, 3-methoxy, 3-methoxypyrocatechol, pyrocatechol, 3-methoxy, 2,3-dihydroxyanisole, pyrogallol 1-methyl ether, 1,2-dihydroxy-3-methoxybenzene, 6-methoxycatechol, pyrogallol 1-monomethyl ether, 3-methoxy-1,2-benzenediol
COC1=CC=CC(=C1O)O
140.138
140.14
≥98.0% (GC)
Safety and Handling

Safety and Handling

RTECSNumber : CZ9002750

TSCA : No

Recommended Storage : Refrigerator